Merck
  • Nucleic acid binding proteins affect the subcellular distribution of phosphorothioate antisense oligonucleotides.
Nucleic acid binding proteins affect the subcellular distribution of phosphorothioate antisense oligonucleotides.
Nucleic acids research (2017-10-05)
Jeffrey K Bailey, Wen Shen, Xue-Hai Liang, Stanley T Crooke
ABSTRACT
Antisense oligonucleotides (ASOs) are versatile tools that can regulate multiple steps of RNA biogenesis in cells and living organisms. Significant improvements in delivery, potency, and stability have been achieved through modifications within the oligonucleotide backbone, sugar and heterocycles. However, these modifications can profoundly affect interactions between ASOs and intracellular proteins in ways that are only beginning to be understood. Here, we report that ASOs with specific backbone and sugar modifications can become localized to cytoplasmic ribonucleoprotein granules such as stress granules and those seeded by the aggregation of specific ASO-binding proteins such as FUS/TLS (FUS) and PSF/SFPQ (PSF). Further investigation into the basis for ASO-FUS binding illustrated the importance of ASO backbone and hydrophobic 2' sugar modifications and revealed that the C-terminal region of FUS is sufficient to retain ASOs in cellular foci. Taken together, the results of this study demonstrate that affinities of various nucleic acid binding domains for ASO depend on chemical modifications and further demonstrate how ASO-protein interactions influence the localization of ASOs.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, asymmetric (ASYM24), serum, Upstate®