Merck
Universal (switchable) RAFT agents.
Journal of the American Chemical Society (2009-05-01)
Massimo Benaglia, John Chiefari, Yen K Chong, Graeme Moad, Ezio Rizzardo, San H Thang
ABSTRACT
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being polymerized. Thus, RAFT agents (dithioesters, trithiocarbonates) suitable for controlling polymerization of "more activated" monomers (MAMs; e.g., styrene, acrylates, methacrylates, etc.) tend to inhibit polymerization of "less activated" monomers (LAMs; e.g., vinyl acetate, N-vinylpyrrolidone, etc.). Similarly RAFT agents suitable for polymerizations of LAMs (xanthates, certain dithiocarbamates) tend to give little or poor control over polymerizations of MAMs. We now report a new class of "switchable" RAFT agents, N-(4-pyridinyl)-N-methyldithiocarbamates, that provide excellent control over polymerization of LAMs and, after addition of 1 equiv of a protic or Lewis acid, become effective in controlling polymerization of MAMs, allowing the synthesis of poly(MAM)-block-poly(LAM) with narrow molecular weight distributions.
Sigma-Aldrich
2-Cyano-2-propyl dodecyl trithiocarbonate, 97% (HPLC)
Sigma-Aldrich
Bis(dodecylsulfanylthiocarbonyl) disulfide, ≥95%
Sigma-Aldrich
4-Cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanoic acid, 97% (HPLC)
Sigma-Aldrich
2-Cyano-2-propyl 4-cyanobenzodithioate, 98% (HPLC)
Sigma-Aldrich
Cyanomethyl methyl(phenyl)carbamodithioate, 98% (HPLC)
Sigma-Aldrich
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid
Sigma-Aldrich
Methyl 2-(dodecylthiocarbonothioylthio)-2-methylpropionate, 97% (HPLC)
Sigma-Aldrich
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid N-hydroxysuccinimide ester, 98% (HPLC)
Sigma-Aldrich
N,N′-Dimethyl N,N′-di(4-pyridinyl)thiuram disulfide
Sigma-Aldrich
Ethyl 2-(phenylcarbonothioylthio)-2-phenylacetate, 98%
Sigma-Aldrich
4-Cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanol
Sigma-Aldrich
Poly(ethylene glycol) methyl ether 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanoate, average Mn 10,000
Sigma-Aldrich
Pentaerythritol tetrakis[2-(dodecylthiocarbonothioylthio)-2-methylpropionate], 97% (HPLC)
Sigma-Aldrich
Cyanomethyl dodecyl trithiocarbonate, 98% (HPLC)
Sigma-Aldrich
2-Cyanopropan-2-yl N-methyl-N-(pyridin-4-yl)carbamodithioate, 97% (HPLC)
Sigma-Aldrich
Bis(thiobenzoyl) disulfide, >95% (HPLC)
Sigma-Aldrich
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid, 98% (HPLC)
Sigma-Aldrich
2-Phenyl-2-propyl benzodithioate, 99% (HPLC)
Sigma-Aldrich
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid N-succinimidyl ester
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Poly(ethylene glycol) bis[2-(dodecylthiocarbonothioylthio)-2-methylpropionate], average Mn 10,800
Sigma-Aldrich
2-(Dodecylthiocarbonothioylthio)propionic acid, 97%
Sigma-Aldrich
2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid 3-azido-1-propanol ester, 98% (HPLC)
Sigma-Aldrich
1,1,1-Tris[(dodecylthiocarbonothioylthio)-2-methylpropionate]ethane, 98% (HPLC)
Sigma-Aldrich
Poly(ethylene glycol) 4-cyano-4-(phenylcarbonothioylthio)pentanoate, average Mn 2,000
Sigma-Aldrich
Poly(ethylene glycol) 4-cyano-4-(phenylcarbonothioylthio)pentanoate, average Mn 10,000