Merck
  • Gut colonization with extended-spectrum β-lactamase-producing Enterobacteriaceae may increase disease activity in biologic-naive outpatients with ulcerative colitis: an interim analysis.
Gut colonization with extended-spectrum β-lactamase-producing Enterobacteriaceae may increase disease activity in biologic-naive outpatients with ulcerative colitis: an interim analysis.
European journal of gastroenterology & hepatology (2017-10-28)
Vita Skuja, Aleksejs Derovs, Katrina Pekarska, Dace Rudzite, Elvira Lavrinovica, Linda Piekuse, Inga Kempa, Zane Straume, Jelena Eglite, Aivars Lejnieks, Angelika Krumina, Rami Eliakim
ABSTRACT
Certain Enterobacteriaceae strains have been associated with the development of ulcerative colitis (UC). Extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae are the most commonly found multi-drug-resistant (MDR) bacteria colonizing the gut in UC patients and might trigger a more severe disease activity in UC patients. The aim of this study was to evaluate whether disease activity is higher in UC patients with gut colonization with ESBL-producing Enterobacteriaceae. A cross-sectional, pilot study was carried out in a tertiary medical center in Latvia. Demographic data were collected; UC disease activity and extent were evaluated according to the full Mayo score, Montreal classification, and adapted Truelove and Witt's index. Rectal swabs with fecal biomaterial were collected, ESBL-producing Enterobacteriaceae were isolated, and bacterial plasmid genes responsible for ESBL production, blaCTX-M, blaTEM, and blaSHV, were detected. UC disease activity was compared in patients with and without gut colonization with ESBL-producing Enterobacteriaceae. A total of 65 patients with UC were included in the initial analysis. Gut colonization with ESBL-producing Enterobacteriaceae was found in seven (11%) patients - mostly Escherichia coli [5 (71%)] containing the blaCTX-M bacterial plasmid gene. Patients with gut colonization with ESBL-producing Enterobacteriaceae had more severe disease compared with patients without gut colonization according to the full Mayo score (5.86 vs. 3.40; P=0.015), Montreal classification (moderate disease vs. clinical remission; P=0.031), and adapted Truelove and Witt's index (moderate disease vs. mild disease; P=0.008). Gut colonization with ESBL-producing Enterobacteriaceae may increase UC disease activity. Further research is needed to analyze the possible confounding factors that could contribute toward this outcome.
Product Description
Supelco
Cefpodoxime, VETRANAL®, analytical standard