Merck
  • Synthesis of Complex Druglike Molecules by the Use of Highly Functionalized Bench-Stable Organozinc Reagents.
Synthesis of Complex Druglike Molecules by the Use of Highly Functionalized Bench-Stable Organozinc Reagents.
Angewandte Chemie (International ed. in English) (2016-10-22)
Thomas J Greshock, Keith P Moore, Ray T McClain, Ana Bellomo, Cheol K Chung, Spencer D Dreher, Peter S Kutchukian, Zhengwei Peng, Ian W Davies, Petr Vachal, Mario Ellwart, Sophia M Manolikakes, Paul Knochel, Philippe G Nantermet
ABSTRACT
The reactivity of a representative set of 17 organozinc pivalates with 18 polyfunctional druglike electrophiles (informers) in Negishi cross-coupling reactions was evaluated by high-throughput experimentation protocols. The high-fidelity scaleup of successful reactions in parallel enabled the isolation of sufficient material for biological testing, thus demonstrating the high value of these new solid zinc reagents in a drug-discovery setting and potentially for many other applications in chemistry. Principal component analysis (PCA) clearly defined the independent roles of the zincates and the informers toward druggable-space coverage.
Sigma-Aldrich
QPhos Pd G3 ChemBeads
Sigma-Aldrich
3-((5-Bromopyridin-2-yl)oxy)-5,5-dimethyl-4-(4-(methylsulfonyl)phenyl)furan-2(5H)-one
Sigma-Aldrich
tert-Butyl (R)-3-(2-acetamidopropan-2-yl)-6-chloro-5-methyl-2,3-dihydrospiro[indene-1,4′-piperidine]-1′-carboxylate
Sigma-Aldrich
2-(5-(3-(4-(2-Bromo-5-fluorophenoxy)piperidin-1-yl)isoxazol-5-yl)-2H-tetrazol-2-yl)acetic acid
Sigma-Aldrich
Methyl 3-(3-(tert-butylthio)-1-(4-chlorobenzyl)-5-(quinolin-2-ylmethoxy)-1H-indol-2-yl)-2,2-dimethylpropanoate
Sigma-Aldrich
1-(Tert-butyl) 2-methyl (2S,4R)-4-((4-bromo-7-fluoroisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate
Sigma-Aldrich
Benzyl (S)-2-(5-bromo-1H-indole-3-carbonyl)pyrrolidine-1-carboxylate
Sigma-Aldrich
Ethyl 4-(3-bromo-8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
Sigma-Aldrich
1-Benzyl 2-methyl (2S,4R)-4-((4-bromoisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate
Sigma-Aldrich
QPhos Pd G3