Merck
  • Chemistry informer libraries: a chemoinformatics enabled approach to evaluate and advance synthetic methods.
Chemistry informer libraries: a chemoinformatics enabled approach to evaluate and advance synthetic methods.
Chemical science (2016-04-21)
Peter S Kutchukian, James F Dropinski, Kevin D Dykstra, Bing Li, Daniel A DiRocco, Eric C Streckfuss, Louis-Charles Campeau, Tim Cernak, Petr Vachal, Ian W Davies, Shane W Krska, Spencer D Dreher
ABSTRACT
Major new advances in synthetic chemistry methods are typically reported using simple, non-standardized reaction substrates, and reaction failures are rarely documented. This makes the evaluation and choice of a synthetic method difficult. We report a standardized complex molecule diagnostic approach using collections of relevant drug-like molecules which we call chemistry informer libraries. With this approach, all chemistry results, successes and failures, can be documented to compare and evolve synthetic methods. To aid in the visualization of chemistry results in drug-like physicochemical space we have used an informatics methodology termed principal component analysis. We have validated this method using palladium- and copper-catalyzed reactions, including Suzuki-Miyaura, cyanation and Buchwald-Hartwig amination.
Sigma-Aldrich
7-(3-Bromo-4-methoxybenzyl)-1-ethyl-8-(((1R,2R)-2-hydroxycyclopentyl)amino)-3-(2-hydroxyethyl)-3,7-dihydro-1H-purine-2,6-dione
Sigma-Aldrich
N-(tert-Butyl)-4′-((6-iodo-4-oxo-2-propylquinazolin-3(4H)-yl)methyl)-[1,1′-biphenyl]-2-sulfonamide
Sigma-Aldrich
tert-Butyl (R)-3-(2-acetamidopropan-2-yl)-6-chloro-5-methyl-2,3-dihydrospiro[indene-1,4′-piperidine]-1′-carboxylate
Sigma-Aldrich
tert-Butyl (S,Z)-(4-(4-bromothiophen-2-yl)-1,4-dimethyl-6-oxotetrahydropyrimidin-2(1H)-ylidene)carbamate
Sigma-Aldrich
Methyl 2-(9-bromo-2,3-dioxo-2,3,6,7-tetrahydro-1H,5H-pyrido[1,2,3-de]quinoxalin-5-yl)acetate, ≥90%
Sigma-Aldrich
3-((5-Bromopyridin-2-yl)oxy)-5,5-dimethyl-4-(4-(methylsulfonyl)phenyl)furan-2(5H)-one
Sigma-Aldrich
2-(5-(3-(4-(2-Bromo-5-fluorophenoxy)piperidin-1-yl)isoxazol-5-yl)-2H-tetrazol-2-yl)acetic acid
Sigma-Aldrich
5-Bromo-N-(4-fluorobenzyl)-8-hydroxy-1,6-naphthyridine-7-carboxamide, ≥90%
Sigma-Aldrich
Methyl 3-(3-(tert-butylthio)-1-(4-chlorobenzyl)-5-(quinolin-2-ylmethoxy)-1H-indol-2-yl)-2,2-dimethylpropanoate
Sigma-Aldrich
4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
Sigma-Aldrich
Benzyl (S)-2-(5-bromo-1H-indole-3-carbonyl)pyrrolidine-1-carboxylate
Sigma-Aldrich
(R)-2-(5-Bromo-4-(4-chlorobenzyl)-7-fluoro-1,2,3,4-tetrahydrocyclopenta[b]indol-3-yl)acetic acid
Sigma-Aldrich
Ethyl 4-(3-bromo-8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
Sigma-Aldrich
1-Benzyl 2-methyl (2S,4R)-4-((4-bromoisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate
Sigma-Aldrich
(R)-5-((1H-1,2,3-Triazol-1-yl)methyl)-3-(3-fluoro-4-iodophenyl)oxazolidin-2-one
Sigma-Aldrich
1-(Tert-butyl) 2-methyl (2S,4R)-4-((4-bromo-7-fluoroisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate