Merck
  • CDK4 Phosphorylates AMPKα2 to Inhibit Its Activity and Repress Fatty Acid Oxidation.
CDK4 Phosphorylates AMPKα2 to Inhibit Its Activity and Repress Fatty Acid Oxidation.
Molecular cell (2017-10-21)
Isabel C Lopez-Mejia, Sylviane Lagarrigue, Albert Giralt, Laia Martinez-Carreres, Nadège Zanou, Pierre-Damien Denechaud, Judit Castillo-Armengol, Carine Chavey, Meritxell Orpinell, Brigitte Delacuisine, Anita Nasrallah, Caterina Collodet, Lianjun Zhang, Benoît Viollet, D Grahame Hardie, Lluis Fajas
ABSTRACT
The roles of CDK4 in the cell cycle have been extensively studied, but less is known about the mechanisms underlying the metabolic regulation by CDK4. Here, we report that CDK4 promotes anaerobic glycolysis and represses fatty acid oxidation in mouse embryonic fibroblasts (MEFs) by targeting the AMP-activated protein kinase (AMPK). We also show that fatty acid oxidation (FAO) is specifically induced by AMPK complexes containing the α2 subunit. Moreover, we report that CDK4 represses FAO through direct phosphorylation and inhibition of AMPKα2. The expression of non-phosphorylatable AMPKα2 mutants, or the use of a CDK4 inhibitor, increased FAO rates in MEFs and myotubes. In addition, Cdk4
Sigma-Aldrich
RB Active human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture