Merck
  • A new photocrosslinkable polycaprolactone-based ink for three-dimensional inkjet printing.
A new photocrosslinkable polycaprolactone-based ink for three-dimensional inkjet printing.
Journal of biomedical materials research. Part B, Applied biomaterials (2016-05-15)
Yinfeng He, Christopher J Tuck, Elisabetta Prina, Sam Kilsby, Steven D R Christie, Stephen Edmondson, Richard J M Hague, Felicity R A J Rose, Ricky D Wildman
ABSTRACT
A new type of photocrosslinkable polycaprolactone (PCL) based ink that is suitable for three-dimensional (3D) inkjet printing has been developed. Photocrosslinkable Polycaprolactone dimethylacrylate (PCLDMA) was synthesized and mixed with poly(ethylene glycol) diacrylate (PEGDA) to prepare an ink with a suitable viscosity for inkjet printing. The ink performance under different printing environments, initiator concentrations, and post processes was studied. This showed that a nitrogen atmosphere during printing was beneficial for curing and material property optimization, as well as improving the quality of structures produced. A simple structure, built in the z-direction, demonstrated the potential for this material for the production of 3D printed objects. Cell tests were carried out to investigate the biocompatibility of the developed ink. © 2016 The Authors Journal of Biomedical Materials Research Part B: Applied Biomaterials Published by Wiley Periodicals, Inc. J Biomed Mater Res Part B: Appl Biomater, 105B: 1645-1657, 2017.
Sigma-Aldrich
2,4-Diethyl-9H-thioxanthen-9-one, 98%
Sigma-Aldrich
Polycaprolactone dimethacrylate, average Mn 10,000
Sigma-Aldrich
Polycaprolactone diacrylate, average Mn 5,000
Sigma-Aldrich
Polycaprolactone dimethacrylate, average Mn 5,000
Sigma-Aldrich
Polycaprolactone diacrylate, average Mn 10,000