Merck
  • Cobalt Phosphonates as Precatalysts for Water Oxidation: Role of Pore Size in Catalysis.
Cobalt Phosphonates as Precatalysts for Water Oxidation: Role of Pore Size in Catalysis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-07-01)
Jony Saha, Debarati Roy Chowdhury, Priyajit Jash, Amit Paul
ABSTRACT
We report a simple approach for the synthesis of cobalt phosphonate (CoOP) nanocages with two distinct types of pore diameters by utilizing a novel tetra-constituent assembly of CoCl
Sigma-Aldrich
Nitrilotris(methylene)triphosphonic acid solution, 50 wt. % in H2O
Sigma-Aldrich
N-(3-Methoxypropyl)acrylamide, contains MEHQ as inhibitor, 95%