Merck
Myc increases self-renewal in neural progenitor cells through Miz-1.
Journal of cell science (2008-11-13)
Laura Kerosuo, Katja Piltti, Heli Fox, Alexandre Angers-Loustau, Valtteri Häyry, Martin Eilers, Hannu Sariola, Kirmo Wartiovaara
ABSTRACT
The mechanisms underlying the decision of a stem or progenitor cell to either self-renew or differentiate are incompletely understood. To address the role of Myc in this process, we expressed different forms of the proto-oncogene Myc in multipotent neural progenitor cells (NPCs) using retroviral transduction. Expression of Myc in neurospheres increased the proportion of self-renewing cells fivefold, and 1% of the Myc-overexpressing cells, but none of the control cells, retained self-renewal capacity even under differentiation-inducing conditions. A Myc mutant (MycV394D) deficient in binding to Miz-1, did not increase the percentage of self-renewing cells but was able to stimulate proliferation of NPCs as efficiently as wild-type Myc, indicating that these two cellular phenomena are regulated by at least partially different pathways. Our results suggest that Myc, through Miz-1, enhances self-renewal of NPCs and influences the way progenitor cells react to the environmental cues that normally dictate the cellular identity of tissues containing self-renewing cells.
Sigma-Aldrich
Saponin from Quillaja sp., Sapogenin content 20-35 %
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000