Merck
  • Evaluation of White Sesame Seed Oil on Glucose Control and Biomarkers of Hepatic, Cardiac, and Renal Functions in Male Sprague-Dawley Rats with Chemically Induced Diabetes.
Evaluation of White Sesame Seed Oil on Glucose Control and Biomarkers of Hepatic, Cardiac, and Renal Functions in Male Sprague-Dawley Rats with Chemically Induced Diabetes.
Journal of medicinal food (2017-03-24)
Farhan Aslam, Sanaullah Iqbal, Muhammad Nasir, Aftab Ahmad Anjum, Pamela Swan, Karen Sweazea
ABSTRACT
White sesame seed oil (WSSO) has been used in cooking and food preparations for centuries. It has many purported health benefits and may be a promising nutraceutical. The primary purpose of this study was to examine the effects of WSSO on fasting blood glucose (GLU) and insulin (INS) in male Sprague-Dawley rats with chemically induced diabetes. A secondary aim was to explore other hematological biomarkers of hepatic, cardiac, and renal function. Sixty-three male Sprague-Dawley rats were randomized into standard diet groups, normal control (NCON) (n = 21) and diabetic control (DCON) (n = 21), and a diabetic sesame oil (DSO) (n = 21) group, which were fed a diet containing 12% WSSO. Blood samples were analyzed at 0, 30, and 60 days. Differences between groups and across days were assessed with two-way repeated measures analysis of variance. At baseline, GLU and INS were similar in both diabetic groups, mean 248.4 ± 2.8 mg/dL and mean 23.4 ± 0.4 μU/mL, respectively. At 60 days, GLU was significantly (P < .05) higher in DCON (298.0 ± 2.3 mg/dL) compared with DSO (202.1 ± 1.0 mg/dL). INS showed similar favorable trends after WSSO supplementation. Consumption of WSSO significantly improved glucose control and other biomarkers of hepatic stress, as well as cardiac and renal health. WSSO may be a viable functional food to help reduce the detrimental effects of diabetes.
Sigma-Aldrich
Alkaline Phosphatase Diethanolamine Activity Kit, sufficient for 100 tests
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Urea Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Mouse Ins1 / Insulin-1 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
BCG Albumin Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
Creatine Kinase Activity Assay Kit, sufficient for 100 tests (Ultraviolet)
Sigma-Aldrich
γ-Globulins from mouse, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Total Protein Kit, Micro Lowry, Peterson′s Modification