Merck
  • Infection-derived lipids elicit an immune deficiency circuit in arthropods.
Infection-derived lipids elicit an immune deficiency circuit in arthropods.
Nature communications (2017-02-15)
Dana K Shaw, Xiaowei Wang, Lindsey J Brown, Adela S Oliva Chávez, Kathryn E Reif, Alexis A Smith, Alison J Scott, Erin E McClure, Vishant M Boradia, Holly L Hammond, Eric J Sundberg, Greg A Snyder, Lei Liu, Kathleen DePonte, Margarita Villar, Massaro W Ueti, José de la Fuente, Robert K Ernst, Utpal Pal, Erol Fikrig, Joao H F Pedra
ABSTRACT
The insect immune deficiency (IMD) pathway resembles the tumour necrosis factor receptor network in mammals and senses diaminopimelic-type peptidoglycans present in Gram-negative bacteria. Whether unidentified chemical moieties activate the IMD signalling cascade remains unknown. Here, we show that infection-derived lipids 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol (POPG) and 1-palmitoyl-2-oleoyl diacylglycerol (PODAG) stimulate the IMD pathway of ticks. The tick IMD network protects against colonization by three distinct bacteria, that is the Lyme disease spirochete Borrelia burgdorferi and the rickettsial agents Anaplasma phagocytophilum and A. marginale. Cell signalling ensues in the absence of transmembrane peptidoglycan recognition proteins and the adaptor molecules Fas-associated protein with a death domain (FADD) and IMD. Conversely, biochemical interactions occur between x-linked inhibitor of apoptosis protein (XIAP), an E3 ubiquitin ligase, and the E2 conjugating enzyme Bendless. We propose the existence of two functionally distinct IMD networks, one in insects and another in ticks.
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, chloroform
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 DG, 1-palmitoyl-2-oleoyl-sn-glycerol, neat oil
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
14:0-16:0 PC, 1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0-16:0 PC, 1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine, powder
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Ubiquitin (pan) Antibody, UbiSite ,clone 2G7B8, clone 2G7B8, from mouse
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution