Merck
  • Directed differentiation of postnatal hippocampal neural stem cells generates nuclear receptor related‑1 protein‑ and tyrosine hydroxylase‑expressing cells.
Directed differentiation of postnatal hippocampal neural stem cells generates nuclear receptor related‑1 protein‑ and tyrosine hydroxylase‑expressing cells.
Molecular medicine reports (2016-07-20)
Yinxiu Ding, Zixin Zhang, Jiangbo Ma, Hechun Xia, Yin Wang, Yinming Liu, Quanrui Ma, Tao Sun, Juan Liu
ABSTRACT
Parkinson's disease (PD) is a severe neurodegenerative disorder. Although the detailed underlying molecular mechanism remains to be elucidated, the major pathological feature of PD is the loss of dopaminergic (DA) neurons of the substantia nigra. The use of donor stem cells to replace DA neurons may be a key breakthrough in the treatment of PD. In the present study, the growth kinetics of hippocampal neural stem cells (Hip‑NSCs) isolated from postnatal mice and cultured in vitro were observed, specifically the generation of cells expressing DA neuronal markers nuclear receptor related‑1 protein (Nurr1) and tyrosine hydroxylase (TH). It was revealed that Hip‑NSCs differentiated primarily into astrocytes when cultured in serum‑containing medium. However, in low serum conditions, the number of βIII tubulin‑positive neurons increased markedly. The proportion of Nurr1‑positive cells and TH‑positive neurons, significantly increased with increasing duration of directed differentiation of Hip‑NSCs (P=0.0187 and 0.0254, respectively). The results of the present study reveal that Hip‑NSCs may be induced to differentiate in vitro into neurons expressing Nurr1 and TH, known to be critical regulators of DA neuronal fate. Additionally, their expression may be necessary to facilitate neuronal maturation in vitro. These data suggest that Hip‑NSCs may serve as a source of DA neurons for cell therapy in patients diagnosed with PD.
Sigma-Aldrich
Anti-GFAP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tyrosine Hydroxylase antibody produced in mouse, clone TH-16, ascites fluid
Sigma-Aldrich
Anti-Tubulin β-III antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Tubulin Isotype III antibody, Mouse monoclonal, clone SDL.3D10, purified from hybridoma cell culture