Merck
  • Ceramide triggers metacaspase-independent mitochondrial cell death in yeast.
Ceramide triggers metacaspase-independent mitochondrial cell death in yeast.
Cell cycle (Georgetown, Tex.) (2011-11-11)
Didac Carmona-Gutierrez, Angela Reisenbichler, Petra Heimbucher, Maria A Bauer, Ralf J Braun, Christoph Ruckenstuhl, Sabrina Büttner, Tobias Eisenberg, Patrick Rockenfeller, Kai-Uwe Fröhlich, Guido Kroemer, Frank Madeo
ABSTRACT
The activation of ceramide-generating enzymes, the blockade of ceramide degradation, or the addition of ceramide analogues can trigger apoptosis or necrosis in human cancer cells. Moreover, endogenous ceramide plays a decisive role in the killing of neoplastic cells by conventional anticancer chemotherapeutics. Here, we explored the possibility that membrane-permeable C2-ceramide might kill budding yeast (Saccharomyces cerevisiae) cells under fermentative conditions, where they exhibit rapid proliferation and a Warburg-like metabolism that is reminiscent of cancer cells. C2-ceramide efficiently induced the generation of reactive oxygen species (ROS), as well as apoptotic and necrotic cell death, and this effect was not influenced by deletion of the sole yeast metacaspase. However, C2-ceramide largely failed to cause ROS hypergeneration and cell death upon deletion of the mitochondrial genome. Thus, mitochondrial function is strictly required for C2-ceramide-induced yeast lethality. Accordingly, mitochondria from C2-ceramide-treated yeast cells exhibited major morphological alterations including organelle fragmentation and aggregation. Altogether, our results point to a pivotal role of mitochondria in ceramide-induced yeast cell death.
Sigma-Aldrich
Dihydroceramide C2, ≥97% (TLC), solid
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection