Merck
Cell-laden microengineered gelatin methacrylate hydrogels.
Biomaterials (2010-04-27)
Jason W Nichol, Sandeep T Koshy, Hojae Bae, Chang M Hwang, Seda Yamanlar, Ali Khademhosseini
ABSTRACT
The cellular microenvironment plays an integral role in improving the function of microengineered tissues. Control of the microarchitecture in engineered tissues can be achieved through photopatterning of cell-laden hydrogels. However, despite high pattern fidelity of photopolymerizable hydrogels, many such materials are not cell-responsive and have limited biodegradability. Here, we demonstrate gelatin methacrylate (GelMA) as an inexpensive, cell-responsive hydrogel platform for creating cell-laden microtissues and microfluidic devices. Cells readily bound to, proliferated, elongated, and migrated both when seeded on micropatterned GelMA substrates as well as when encapsulated in microfabricated GelMA hydrogels. The hydration and mechanical properties of GelMA were demonstrated to be tunable for various applications through modification of the methacrylation degree and gel concentration. The pattern fidelity and resolution of GelMA were high and it could be patterned to create perfusable microfluidic channels. Furthermore, GelMA micropatterns could be used to create cellular micropatterns for in vitro cell studies or 3D microtissue fabrication. These data suggest that GelMA hydrogels could be useful for creating complex, cell-responsive microtissues, such as endothelialized microvasculature, or for other applications that require cell-responsive microengineered hydrogels.
Sigma-Aldrich
Low endotoxin GelMA, mol wt 95 kDa, degree of substitution 60%
Sigma-Aldrich
Gelatin methacryloyl, gel strength 90-110 g Bloom, degree of substitution 60%
Sigma-Aldrich
Gelatin methacryloyl, gel strength 170-195 g Bloom, degree of substitution: 60%
Sigma-Aldrich
Gelatin methacryloyl, gel strength 300 g Bloom, degree of substitution 60%
Sigma-Aldrich
Low endotoxin GelMA, gel strength 300 (Bloom), degree of substitution 80%, GelMA Type B
Sigma-Aldrich
Low endotoxin GelMA, bloom 300, Type A, degree of substitution 80%
Sigma-Aldrich
Gelatin methacryloyl, gel strength 300 g Bloom, 80% degree of substitution
Sigma-Aldrich
Gelatin methacryloyl, gel strength 300 g Bloom, degree of substitution 40%
Sigma-Aldrich
Low endotoxin GelMA solution, gel strength 300 (bloom), degree of substitution 80%, 0.2 μm, sterile-filtered, GelMA Type B
Sigma-Aldrich
TissueFab® Discrete GelMA, 90-110 bloom, 50% degree of substitution
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 80% degree of substitution
Sigma-Aldrich
TissueFab® Discrete GelMA, 300 bloom, 50% degree of substitution