Merck
  • Phosphorus-containing inhibitors of HMG-CoA reductase. 2. Synthesis and biological activities of a series of substituted pyridines containing a hydroxyphosphinyl moiety.
Phosphorus-containing inhibitors of HMG-CoA reductase. 2. Synthesis and biological activities of a series of substituted pyridines containing a hydroxyphosphinyl moiety.
Journal of medicinal chemistry (1991-09-11)
J A Robl, L A Duncan, J Pluscec, D S Karanewsky, E M Gordon, C P Ciosek, L C Rich, V C Dehmel, D A Slusarchyk, T W Harrity
ABSTRACT
A series of 2,3,4,(5),6-substituted pyridines containing a hydroxyphosphinyl functionally have been prepared and were evaluated for their ability to inhibit the enzyme HMG-CoA reductase. Systematic substitution of both R1-R4 and X-Y led to compounds of type 3-6 with in vitro potency greater than that of mevinolin (Na salt).
Sigma-Aldrich
Bromotrimethylsilane, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl(chloro)diphenylsilane, 98%
Pricing and availability is not currently available.