Merck
  • Endonuclease G initiates DNA rearrangements at the MLL breakpoint cluster upon replication stress.
Endonuclease G initiates DNA rearrangements at the MLL breakpoint cluster upon replication stress.
Oncogene (2014-08-19)
B Gole, C Baumann, E Mian, C I Ireno, L Wiesmüller
ABSTRACT
MLL (myeloid/lymphoid or mixed-lineage leukemia) rearrangements are frequent in therapy-related and childhood acute leukemia, and are associated with poor prognosis. The majority of the rearrangements fall within a 7.3-kb MLL breakpoint cluster region (MLLbcr), particularly in a 0.4-kb hotspot at the intron11-exon12 boundary. The underlying mechanisms are poorly understood, though multiple pathways including early apoptotic signaling, accompanied by high-order DNA fragmentation, have been implicated. We introduced the MLLbcr hotspot in an EGFP-based recombination reporter system and demonstrated enhancement of both spontaneous and genotoxic treatment-induced DNA recombination by the MLLbcr in various human cell types. We identified Endonuclease G (EndoG), an apoptotic nuclease, as an essential factor for MLLbcr-specific DNA recombination after induction of replication stress. We provide evidence for replication stress-induced nuclear accumulation of EndoG, DNA binding by EndoG as well as cleavage of the chromosomal MLLbcr locus in a manner requiring EndoG. We demonstrate additional dependency of MLLbcr breakage on ATM signaling to histone H2B monoubiquitinase RNF20, involved in chromatin relaxation. Altogether our findings provide a novel mechanism underlying MLLbcr destabilization in the cells of origin of leukemogenesis, with replication stress-activated, EndoG-mediated cleavage at the MLLbcr, which may serve resolution of the stalled forks via recombination repair, however, also permits MLL rearrangements.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-DFF40 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Z-Leu-Leu-Norvalinal, ≥90% (HPLC), powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder