Merck
  • Purification and characterization of antioxidant peptides from enzymatically hydrolyzed chicken egg white.
Purification and characterization of antioxidant peptides from enzymatically hydrolyzed chicken egg white.
Food chemistry (2015-06-05)
Chamila Nimalaratne, Nandika Bandara, Jianping Wu
ABSTRACT
Egg white is considered as a rich source of high quality proteins with various bioactive peptide fractions. Enzymatic hydrolysis of proteins can be used to release bioactive fractions and different enzymes have different abilities in releasing such bioactive fractions depending on the enzyme's site of activity on a protein. In this study, several proteases were examined for their ability to release antioxidant peptides from hen egg white and protease P was selected based on the antioxidant activity and the digestion yield of the crude protein hydrolysate. A combination of several purification steps including ultrafiltration with low molecular weight cut-off membranes, cation exchange chromatography and reversed phase high performance liquid chromatography was used to purify 'protease P egg white hydrolysate'. Sixteen antioxidant peptides, which were derived from ovalbumin, ovotransferrin and cystatin were isolated from the most active fractions. Amino acid sequences of those peptides were determined using LC-MS/MS. Oxygen radical absorbance capacity (ORAC) values of selected short chain peptides were determined using synthetic peptides. Two peptides AEERYP and DEDTQAMP (Ala-Glu-Glu-Arg-Tyr-Pro and Asp-Glu-Asp-Thr-Gln-Ala-Met-Pro) showed the highest ORAC values. The results from this study indicate that egg white is rich in antioxidant peptides which can be used as a potential source for preparing bioactive ingredients using enzymatic hydrolysis followed by purification techniques.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.