Merck
  • Genericity of confined chemical garden patterns with regard to changes in the reactants.
Genericity of confined chemical garden patterns with regard to changes in the reactants.
Physical chemistry chemical physics : PCCP (2015-04-25)
Florence Haudin, V Brasiliense, Julyan H E Cartwright, Fabian Brau, A De Wit
ABSTRACT
The growth of chemical gardens is studied experimentally in a horizontal confined geometry when a solution of metallic salt is injected into an alkaline solution at a fixed flow rate. Various precipitate patterns are observed-spirals, flowers, worms or filaments-depending on the reactant concentrations. In order to determine the relative importance of the chemical nature of the reactants and physical processes in the pattern selection, we compare the structures obtained by performing the same experiment using different pairs of reactants of varying concentrations with cations of calcium, cobalt, copper, and nickel, and anions of silicate and carbonate. We show that although the transition zones between different patterns are not sharply defined, the morphological phase diagrams are similar in the various cases. We deduce that the nature of the chemical reactants is not a key factor for the pattern selection in the confined chemical gardens studied here and that the observed morphologies are generic patterns for precipitates possessing a given level of cohesiveness when grown under certain flow conditions.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis