Merck
  • Evaluation of different methods for immobilization of Candida antarctica lipase B (CalB lipase) in polyurethane foam and its application in the production of geranyl propionate.
Evaluation of different methods for immobilization of Candida antarctica lipase B (CalB lipase) in polyurethane foam and its application in the production of geranyl propionate.
Bioprocess and biosystems engineering (2015-06-04)
Gabrieli Nicoletti, Eliane P Cipolatti, Alexsandra Valério, NatáliaThaisa Gamba Carbonera, Nicole Spillere Soares, Eron Theilacker, Jorge L Ninow, Débora de Oliveira
ABSTRACT
With the aim of studying the best method for the interaction of polyurethane (PU) foam and Candida antarctica lipase B, different methods of CalB immobilization were studied: adsorption (PU-ADS), bond (using polyethyleneimine) (PU-PEI), ionic adsorption by PEI with cross-linking with glutaraldehyde (PU-PEI-GA) and entrapment (PU). The characterization of immobilized enzyme derivatives was performed by apparent density and Fourier transform infrared spectroscopy. The free enzyme and enzyme preparations were evaluated at different pH values and temperatures. The highest enzyme activity was obtained using the PU method (5.52 U/g). The methods that stood out to compare the stabilities and kinetic parameters were the PU and PU-ADS. Conversions of 83.5 and 95.9 % for PU and PU-ADS derivatives were obtained, in 24 h reaction, using citronella oil and propionic acid as substrates.
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Geranyl propionate, >95%, FCC, FG