Merck
  • Surface-Induced Dissociation Mass Spectra as a Tool for Distinguishing Different Structural Forms of Gas-Phase Multimeric Protein Complexes.
Surface-Induced Dissociation Mass Spectra as a Tool for Distinguishing Different Structural Forms of Gas-Phase Multimeric Protein Complexes.
Analytical chemistry (2015-10-27)
Royston S Quintyn, Mowei Zhou, Jing Yan, Vicki H Wysocki
ABSTRACT
One attractive feature of ion mobility mass spectrometry (IM-MS) lies in its ability to provide experimental collision cross section (CCS) measurements, which can be used to distinguish different conformations that a protein complex may adopt during its gas-phase unfolding. However, CCS values alone give no detailed information on subunit structure within the complex. Consequently, structural characterization typically requires molecular modeling, which can have uncertainties without experimental support. One method of obtaining direct experimental evidence on the structures of these intermediates is utilizing gas-phase activation techniques that can effectively dissociate the complexes into substructures while preserving the native topological information. The most commonly used activation method, collision-induced dissociation (CID) with low-mass target gases, typically leads to unfolding of monomers of a protein complex. Here, we describe a method that couples IM-MS and surface-induced dissociation (SID) to dissociate the source-activated precursors of three model protein complexes: C-reactive protein (CRP), transthyretin (TTR), and concanavalin A (Con A). The results of this study confirm that CID involves the unfolding of the protein complex via several intermediates. More importantly, our experiments also indicate that retention of similar CCS between different intermediates does not guarantee retention of structure. Although CID spectra (at a given collision energy) of source-activated, mass-selected precursors do not distinguish between native-like, collapsed, and expanded forms of a protein complex, dissociation patterns and/or average charge states of monomer products in SID of each of these forms are unique.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%