Merck
  • The application of fuzzy statistics and linear discriminant analysis as criteria for optimizing the preparation of plasma for matrix-assisted laser desorption/ionization mass spectrometry peptide profiling.
The application of fuzzy statistics and linear discriminant analysis as criteria for optimizing the preparation of plasma for matrix-assisted laser desorption/ionization mass spectrometry peptide profiling.
Clinica chimica acta; international journal of clinical chemistry (2015-07-15)
Joanna Hajduk, Jan Matysiak, Piotr Kokot, Piotr Nowicki, Paweł Dereziński, Zenon J Kokot
ABSTRACT
An alternative bioinformatics approach based on fuzzy theory statistics and linear discriminant analysis is proposed for the interpretation of MALDI MS spectra in peptide profiling. When applied, the methodology enables the establishment of a reproducible plasma preparation protocol appropriate for the evaluation of small data sets. The samples were collected from pregnant women affected by gestational diabetes mellitus (GDM), n=18 and control group, n=13. The following pre-treatment sets were tested: pipette tips with C18 stationary phase (ZipTip, Millipore and Omix, Agilent) and magnetic bead-based weak cation exchange chromatography kit (MB WCX, Bruker Daltonics). The spectra were recorded using a MALDI TOF mass spectrometer (UltrafleXtreme, Bruker Daltonics) for a mass range of m/z from 1000 to 10,000. The significant features were selected using the wrapper selection method, and two classification systems were tested: discriminant analysis (DA) and fuzzy inference system (FIS). ClinProTools software was employed to compare the usefulness of the proposed methodology. The study showed that the optimum results for MS spectra were obtained after the use of the ZipTip as pre-treatment method in plasma preparation. Chemometric analysis allowed the differentiation of the GDM group from the control with a high degree of accuracy: 0.7333 (DA) and 0.8065 (FIS).
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%