Merck
  • Engineered catalytic biofilms: Site-specific enzyme immobilization onto E. coli curli nanofibers.
Engineered catalytic biofilms: Site-specific enzyme immobilization onto E. coli curli nanofibers.
Biotechnology and bioengineering (2015-05-08)
Zsofia Botyanszki, Pei Kun R Tay, Peter Q Nguyen, Martin G Nussbaumer, Neel S Joshi
ABSTRACT
Biocatalytic transformations generally rely on purified enzymes or whole cells to perform complex transformations that are used on industrial scale for chemical, drug, and biofuel synthesis, pesticide decontamination, and water purification. However, both of these systems have inherent disadvantages related to the costs associated with enzyme purification, the long-term stability of immobilized enzymes, catalyst recovery, and compatibility with harsh reaction conditions. We developed a novel strategy for producing rationally designed biocatalytic surfaces based on Biofilm Integrated Nanofiber Display (BIND), which exploits the curli system of E. coli to create a functional nanofiber network capable of covalent immobilization of enzymes. This approach is attractive because it is scalable, represents a modular strategy for site-specific enzyme immobilization, and has the potential to stabilize enzymes under denaturing environmental conditions. We site-specifically immobilized a recombinant α-amylase, fused to the SpyCatcher attachment domain, onto E. coli curli fibers displaying complementary SpyTag capture domains. We characterized the effectiveness of this immobilization technique on the biofilms and tested the stability of immobilized α-amylase in unfavorable conditions. This enzyme-modified biofilm maintained its activity when exposed to a wide range of pH and organic solvent conditions. In contrast to other biofilm-based catalysts, which rely on high cellular metabolism, the modified curli-based biofilm remained active even after cell death due to organic solvent exposure. This work lays the foundation for a new and versatile method of using the extracellular polymeric matrix of E. coli for creating novel biocatalytic surfaces.
Sigma-Aldrich
Rosetta Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-6477927-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€444.10
EN300-6477927-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€613.10
EN300-6477927-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€940.70
EN300-6477927-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,194.20
EN300-6477927-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
€2,296.60
EN300-6477927-5G5 g
Ships in 14 days - from Aldrich Partner.
€3,376.90
To order products, please contact your local dealer.
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-6477927-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€444.10
EN300-6477927-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€613.10
EN300-6477927-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€940.70
EN300-6477927-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,194.20
EN300-6477927-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
€2,296.60
EN300-6477927-5G5 g
Ships in 14 days - from Aldrich Partner.
€3,376.90
To order products, please contact your local dealer.