Merck
  • Association of HDL cholesterol efflux capacity with incident coronary heart disease events: a prospective case-control study.
Association of HDL cholesterol efflux capacity with incident coronary heart disease events: a prospective case-control study.
The lancet. Diabetes & endocrinology (2015-05-31)
Danish Saleheen, Robert Scott, Sundas Javad, Wei Zhao, Amrith Rodrigues, Antonino Picataggi, Daniya Lukmanova, Megan L Mucksavage, Robert Luben, Jeffery Billheimer, John J P Kastelein, S Matthijs Boekholdt, Kay-Tee Khaw, Nick Wareham, Daniel J Rader
ABSTRACT
Although HDL cholesterol concentrations are strongly and inversely associated with risk of coronary heart disease, interventions that raise HDL cholesterol do not reduce risk of coronary heart disease. HDL cholesterol efflux capacity-a prototypical measure of HDL function-has been associated with coronary heart disease after adjusting for HDL cholesterol, but its effect on incident coronary heart disease risk is uncertain. We measured cholesterol efflux capacity and assessed its relation with vascular risk factors and incident coronary heart disease events in a nested case-control sample from the prospective EPIC-Norfolk study of 25 639 individuals aged 40-79 years, assessed in 1993-97 and followed up to 2009. We quantified cholesterol efflux capacity in 1745 patients with incident coronary heart disease and 1749 control participants free of any cardiovascular disorders by use of a validated ex-vivo radiotracer assay that involved incubation of cholesterol-labelled J774 macrophages with apoB-depleted serum from study participants. Cholesterol efflux capacity was positively correlated with HDL cholesterol concentration (r=0·40; p<0·0001) and apoA-I concentration (r=0·22; p<0·0001). It was also inversely correlated with type 2 diabetes (r=-0·18; p<0·0001) and positively correlated with alcohol consumption (r=0·12; p<0·0001). In analyses comparing the top and bottom tertiles, cholesterol efflux capacity was significantly and inversely associated with incident coronary heart disease events, independent of age, sex, diabetes, hypertension, smoking and alcohol use, waist:hip ratio, BMI, LDL cholesterol concentration, log-triglycerides, and HDL cholesterol or apoA-I concentrations (odds ratio 0·64, 95% CI 0·51-0·80). After a similar multivariable adjustment the risk of incident coronary heart disease was 0·80 (95% CI 0·70-0·90) for a per-SD change in cholesterol efflux capacity. HDL cholesterol efflux capacity might provide an alternative mechanism for therapeutic modulation of the HDL pathway beyond HDL cholesterol concentration to help reduce risk of coronary heart disease. US National Institutes of Health, UK Medical Research Council, Cancer Research UK.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)