Merck
  • The prediction of the palatability of orally disintegrating tablets by an electronic gustatory system.
The prediction of the palatability of orally disintegrating tablets by an electronic gustatory system.
International journal of pharmaceutics (2015-07-29)
Hideshi Nakamura, Shinya Uchida, Takeshi Sugiura, Noriyuki Namiki
ABSTRACT
In this study, the human gustatory palatability sensation of taste-masked famotidine and amlodipine orally disintegrating tablets (ODTs) was quantitatively predicted by an electronic gustatory system (α-Astree e-Tongue). Furthermore, its use in formulation design was evaluated. The famotidine- and amlodipine-containing ODTs, which were bitter- and highly bitter-tasting, respectively, were prepared using a physical (granules spray-coated with ethyl cellulose) or organoleptic (the addition of a sweetener and a flavor) masking method and combinations thereof. The taste-masking effects of different masking methods on the ODTs were investigated in a human gustatory sensation test. In the test, volunteers scored the overall palatability using a 100mm visual analog scale (VAS). The electronic gustatory system was evaluated using the Euclidean distance (the distance between each drug-containing ODT and its corresponding placebo) and partial least squares (PLS) regression analysis of the sensor response values. A good linear relationship was observed between each ODT's Euclidean distance analysis, PLS regression analysis, and clinical VAS scores. Cross-validation verification of each analysis confirmed the model's predictive power. This study suggests that the α-Astree can quantitatively evaluate physical and organoleptic taste masking and that the palatability of unknown formulations can be predicted by Euclidean distance and PLS regression data analysis.
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 100-200 mesh (75 - 150 μm)
Sigma-Aldrich
Silicic acid, suitable for column chromatography, 60-200 mesh
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)