Merck
  • Machine oil inhibits the osseointegration of orthopaedic implants by impairing osteoblast attachment and spreading.
Machine oil inhibits the osseointegration of orthopaedic implants by impairing osteoblast attachment and spreading.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2015-02-14)
Lindsay A Bonsignore, Victor M Goldberg, Edward M Greenfield
ABSTRACT
The most important factor contributing to short-term and long-term success of cementless total joint arthroplasties is osseointegration. Osseointegration leads to a direct structural and functional connection between living bone and the surface of an implant. Surface contaminants may remain on orthopaedic implants after sterilization procedures and impair osseointegration. For example, specific lots of hip replacement Sulzer Inter-OP(TM) acetabular shells that were associated with impaired osseointegration and early failure rates were found to be contaminated with both bacterial debris and machine oil residues. However, the effect of machine oil on implant integration is unknown. Therefore, the goal of this study was to determine if machine oil inhibits the osseointegration of orthopaedic implants. To test this hypothesis in vivo we used our murine model of osseointegration where titanium alloy implants are implanted into a unicortical pilot hole in the mid-diaphysis of the femur. We found that machine oil inhibited bone-to-implant contact and biomechanical pullout measures. Machine oil on titanium alloy discs inhibited early stages of MC3T3-E1 osteogenesis in vitro such as attachment and spreading. Inhibition of osteoblast attachment and spreading occurred in both areas with and without detectable oil. Osteoblast growth was in turn inhibited on discs with machine oil due to both a decrease in proliferation and an increase in cell death. Later stages of osteogenic differentiation and mineralization on titanium alloy discs were also inhibited. Thus, machine oil can inhibit osseointegration through cell autonomous effects on osteoblast cells. These results support routine testing by manufacturers of machine oil residues on orthopaedic implants.
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%