Merck
  • The polyomavirus middle T-antigen oncogene activates the Hippo pathway tumor suppressor Lats in a Src-dependent manner.
The polyomavirus middle T-antigen oncogene activates the Hippo pathway tumor suppressor Lats in a Src-dependent manner.
Oncogene (2014-11-05)
M Shanzer, I Ricardo-Lax, R Keshet, N Reuven, Y Shaul
ABSTRACT
The polyomavirus middle T antigen (PyMT) is an oncogene that activates the non-receptor tyrosine kinase, c-Src, and physically interacts with Taz (WWTR1). Taz is a pro-oncogenic transcription coactivator of the Tead transcription factors. The Hippo tumor suppressor pathway activates the kinase Lats, which phosphorylates Taz, leading to its nuclear exclusion and blunting Tead coactivation. We found that Taz was required for transformation by PyMT, but counter-intuitively, Taz was exclusively cytoplasmic in the presence of PyMT. We demonstrate that in the presence of PyMT, wild-type Taz was phosphorylated by Lats, in a Src-dependent manner. Consistently, a Lats refractory Taz mutant did not undergo cytoplasmic retention by PyMT. We show that Yap, the Taz paralog, and Shp2 phosphatase were nuclear excluded as well. Our findings describe a noncanonical activation of Lats, and an unprecedented Tead-independent role for Taz and Yap in viral-mediated oncogenesis.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Taz
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wwtr1
Sigma-Aldrich
MISSION® esiRNA, targeting human TAZ
Sigma-Aldrich
MISSION® esiRNA, targeting human WWTR1
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SAFC
HEPES