Merck
  • A wash step at collection of placental biopsies from preeclamptic pregnancies does not adversely affect levels of sFlt-1 or endoglin.
A wash step at collection of placental biopsies from preeclamptic pregnancies does not adversely affect levels of sFlt-1 or endoglin.
Pregnancy hypertension (2015-11-26)
Tu'uhevaha J Kaitu'u-Lino, Natalie J Hannan, Manarangi De Silva, Natalie Binder, Laura Tuohey, Ping Cannon, Stephen Tong
ABSTRACT
There is recent interest for uniform placental collection protocols so laboratories can share samples. However, concerns have been raised a wash step at collection causes significant loss of sFlt-1 and soluble endoglin, among the most studied proteins in placentology. We measured Flt-1 and endoglin mRNA and protein in 10 preeclamptic placentas that were washed, or left unwashed. Reassuringly, there was no significant change in the Flt-1, sFlt-1-e15a or sFlt-1-i13 mRNA or Flt-1 or sFlt-1 protein expression or localization. There was also no change in endoglin mRNA expression or protein localization. Washing preeclamptic placental samples does not alter anti-angiogenic factor expression.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Mini chemical spill kit
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
Sigma-Aldrich
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Adapter for ACE Micro/Mini-Lab® kit, Distilling, joint: ST/NS 14/10
Adapter for ACE Micro/Mini-Lab® kit, Vacuum-takeoff with hose barb, joint: ST/NS 14/10
Adapter for ACE Micro/Mini-Lab® kit, Mini adapter