Merck
  • Syntheses, structures, and (1)H, (13)C{(1)H} and (119)Sn{(1)H} NMR chemical shifts of a family of trimethyltin alkoxide, amide, halide and cyclopentadienyl compounds.
Syntheses, structures, and (1)H, (13)C{(1)H} and (119)Sn{(1)H} NMR chemical shifts of a family of trimethyltin alkoxide, amide, halide and cyclopentadienyl compounds.
Dalton transactions (Cambridge, England : 2003) (2015-08-22)
Alejandro G Lichtscheidl, Michael T Janicke, Brian L Scott, Andrew T Nelson, Jaqueline L Kiplinger
ABSTRACT
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in benzene-d6, toluene-d8, dichloromethane-d2, chloroform-d1, acetonitrile-d3, and tetrahydrofuran-d8 are reported. The X-ray crystal structures of Me3Sn(O-2,6-(t)Bu2C6H3) (), Me3Sn(O-2,6-(i)Pr2C6H3) (), and (Me3Sn)(NH-2,4,6-(t)Bu3C6H2) () are also presented. These compiled data complement existing literature data and ease the characterization of these compounds by routine NMR experiments.
Sigma-Aldrich
Acetonitrile-d3, 96-97 atom % D, D2O 15-20 %
Sigma-Aldrich
Acetonitrile-d3, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Acetonitrile-d3, ≥99.8 atom % D
Sigma-Aldrich
Acetonitrile-d3, "100%", 99.96 atom % D
Sigma-Aldrich
Tetrahydrofuran-d8, ≥99.5 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran-d8, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran-d8, ≥99.5 atom % D
Sigma-Aldrich
Acetonitrile-d3, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Tetrahydrofuran-d8, ≥99.5 atom % D
Sigma-Aldrich
Acetonitrile-d3, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Benzene-D6, anhydrous, ≥99.6 atom % D
Sigma-Aldrich
Benzene-D6, 99.6 atom % D
Sigma-Aldrich
Benzene-D6, "100%", 99.96 atom % D
Sigma-Aldrich
Toluene-d8, "100%", 99.96 atom % D
Sigma-Aldrich
Toluene-d8, 99 atom % D
Sigma-Aldrich
Dichloromethane-d2, "100%", 99.96 atom % D
Sigma-Aldrich
Dichloromethane-d2, 99.9 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Toluene-d8, 99 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Benzene-D6, 99 atom % D
Sigma-Aldrich
Toluene-d8, 99.6 atom % D
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Dichloromethane-d2, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dichloromethane-d2, 99.9 atom % D
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Dichloromethane-d2, 99.5 atom % D
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)