Merck
  • Cytochrome c reacts with cholesterol hydroperoxides to produce lipid- and protein-derived radicals.
Cytochrome c reacts with cholesterol hydroperoxides to produce lipid- and protein-derived radicals.
Biochemistry (2015-04-14)
Thiago C Genaro-Mattos, Raphael F Queiroz, Daniela Cunha, Patricia P Appolinario, Paolo Di Mascio, Iseli L Nantes, Ohara Augusto, Sayuri Miyamoto
ABSTRACT
Lipid peroxidation is a well-known process that has been implicated in many diseases. Recent evidence has shown that mitochondrial cholesterol levels are increased under specific conditions, making it an important target for peroxidation inside the mitochondria. Cholesterol peroxidation generates, as primary products, several hydroperoxides (ChOOH), which can react with transition metals and metalloproteins. In this sense, cytochrome c (CYTC), a heme protein largely found in the mitochondria, becomes a candidate to react with ChOOH. Using CYTC associated with SDS micelles to mimic mitochondrial conditions, we show that ChOOH induces dose-dependent CYTC Soret band bleaching, indicating that it is using ChOOH as a substrate. This reaction leads to protein oligomerization, suggesting the formation of a protein radical that, subsequently, recombines, giving dimers, trimers, and tetramers. EPR experiments confirmed the production of carbon-centered radicals from both protein and lipid in the presence of ChOOH. Similar results were obtained with linoleic acid hydroperoxides (LAOOH). In addition, replacing SDS micelles by cardiolipin-containing liposomes as the mitochondrial mimetic led to similar results with either ChOOH or LAOOH. Importantly, kinetic experiments show that CYTC bleaching is faster with ChOOH than with H2O2, suggesting that these hydroperoxides could be relevant substrates for CYTC peroxidase-like activity in biological media. Altogether, these results show that CYTC induces homolytic cleavage of lipid-derived hydroperoxides, producing lipid and protein radicals.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, ~95%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))