Merck
  • Aerobic Fitness Affects the Exercise Performance Responses to Nitrate Supplementation.
Aerobic Fitness Affects the Exercise Performance Responses to Nitrate Supplementation.
Medicine and science in sports and exercise (2014-11-21)
Simone Porcelli, Matthew Ramaglia, Giuseppe Bellistri, Gaspare Pavei, Lorenzo Pugliese, Michela Montorsi, Letizia Rasica, Mauro Marzorati
ABSTRACT
Dietary nitrate supplementation has been shown to reduce O2 cost of submaximal exercise, improve exercise tolerance, and enhance performance in moderately trained individuals. In contrast, data have been provided that elite athletes do not benefit from nitrate supplementation. The aim of this study was to evaluate the effects of short-term nitrate supplementation on endurance performance in subjects with different levels of aerobic fitness. Twenty-one subjects (mean age, 22.7 ± 1.8 yr) with different aerobic fitness level (V˙O2peak value ranging from 28.2 to 81.7 mL·kg·min) participated in a crossover double-blind placebo-controlled study. Subjects were tested after 6 d of supplementation with either 0.5 l per day of nitrate (5.5 mmol)-containing water (NITR) or nitrate-free water (PLA). Participants performed an incremental running test until exhaustion and four repetitions of 6-min submaximal (approximately 80% of gas exchange threshold) constant load exercise on a motorized treadmill. Moreover, subjects performed a 3-km running time trial on the field. After NITR, a negative correlation between reduction of O2 cost of submaximal exercise and individual aerobic fitness level was observed (r = 0.80; P < 0.0001). A significant inverse correlation was also found between aerobic fitness level and improvement in performance for 3-km time trial after NITR (r = 0.76; P < 0.0001). Additionally, subjects responded differently to dietary nitrate supplementation according to aerobic fitness level with higher-fit subjects showing a lower increase in plasma [NO3] (r = 0.86; P < 0.0001) and [NO2] (r = 0.75; P < 0.0001). The results of the present study suggest that the individual aerobic fitness level affects the ergogenic benefits induced by dietary nitrate supplementation. The optimal nitrate loading regimen required to elevate plasma [NO2] and to enhance performance in elite athletes is different from that of low-fit subjects and requires further studies.
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
O-Acetyl-L-carnitine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis