Merck
  • Quantitative characterization of hydration state and destructuring effect of monosaccharides and disaccharides on water hydrogen bond network.
Quantitative characterization of hydration state and destructuring effect of monosaccharides and disaccharides on water hydrogen bond network.
Carbohydrate research (2015-02-07)
K Shiraga, T Suzuki, N Kondo, J De Baerdemaeker, Y Ogawa
ABSTRACT
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on the hydrogen bond (HB) network. Firstly, the extent of hydration water was determined by the decreased amount of bulk water with picosecond relaxation time that was replaced by that with much longer orientational relaxation time. As a result, we found glucose and trehalose exhibits stronger hydration capacity than fructose and sucrose, respectively, despite of the same number of the hydroxyl groups. For each saccharide, the hydration number tended to decrease with solute concentration. Secondly, the destructuring effect of these saccharide solutes on the HB network of the surrounding bulk water was discussed from the perspective of the fraction of non-hydrogen-bonded (NHB) water isolated from the HB network. We found the fraction of NHB water molecules that are not engaged in the HB network monotonously increased with saccharide concentration, indicating saccharide solutes promote the disruption of the water HB network. However, no noticeable differences were confirmed in the fraction of NHB water between glucose and fructose or between sucrose and trehalose. In contrast to hydration number, the number of NHB water produced by a single saccharide solute was less dependent on solute concentration, and three monosaccharide/disaccharide solutes were found to produce one/two NHB water molecules.
Product Description
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Fructose, European Pharmacopoeia (EP) Reference Standard
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Supelco
Trehalose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trehalose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC)