Merck
  • Origin of the large dispersion of magnetic properties in nanostructured oxides: Fe(x)O/Fe3O4 nanoparticles as a case study.
Origin of the large dispersion of magnetic properties in nanostructured oxides: Fe(x)O/Fe3O4 nanoparticles as a case study.
Nanoscale (2015-01-21)
Marta Estrader, Alberto López-Ortega, Igor V Golosovsky, Sònia Estradé, Alejandro G Roca, German Salazar-Alvarez, Lluís López-Conesa, Dina Tobia, Elin Winkler, José D Ardisson, Waldemar A A Macedo, Andreas Morphis, Marianna Vasilakaki, Kalliopi N Trohidou, Arsen Gukasov, Isabelle Mirebeau, O L Makarova, Roberto D Zysler, Francesca Peiró, Maria Dolors Baró, Lennart Bergström, Josep Nogués
ABSTRACT
The intimate relationship between stoichiometry and physicochemical properties in transition-metal oxides makes them appealing as tunable materials. These features become exacerbated when dealing with nanostructures. However, due to the complexity of nanoscale materials, establishing a distinct relationship between structure-morphology and functionalities is often complicated. In this regard, in the FexO/Fe3O4 system a largely unexplained broad dispersion of magnetic properties has been observed. Here we show, thanks to a comprehensive multi-technique approach, a clear correlation between the magneto-structural properties in large (45 nm) and small (9 nm) FexO/Fe3O4 core/shell nanoparticles that can explain the spread of magnetic behaviors. The results reveal that while the FexO core in the large nanoparticles is antiferromagnetic and has bulk-like stoichiometry and unit-cell parameters, the FexO core in the small particles is highly non-stoichiometric and strained, displaying no significant antiferromagnetism. These results highlight the importance of ample characterization to fully understand the properties of nanostructured metal oxides.
Product Description
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, Selectophore, for ion-selective electrodes, ≥99%
Oleic acid, European Pharmacopoeia (EP) Reference Standard