Merck
  • Influence of ovarian stimulation for IVF/ICSI on the antioxidant defence system and relationship to outcome.
Influence of ovarian stimulation for IVF/ICSI on the antioxidant defence system and relationship to outcome.
Reproductive biomedicine online (2014-05-13)
Simone Palini, Serena Benedetti, Maria Chiara Tagliamonte, Silvia De Stefani, Mariangela Primiterra, Valeria Polli, Patrizia Rocchi, Simona Catalani, Serafina Battistelli, Franco Canestrari, Carlo Bulletti
ABSTRACT
Ovarian stimulation is used with IVF/intracytoplasmic sperm injection (ICSI) cycles to obtain multiple oocytes and improve pregnancy rates; however, it also induces perturbation in the oxidant-antioxidant balance leading to oxidation stress. The present study monitored the plasma antioxidant status in women undergoing a long agonist protocol of ovarian stimulation at three different time points: at baseline (T0), after pituitary suppression (T1) and on the day of oocyte retrieval (T2). The antioxidant composition of follicular fluid samples collected on T2 was also evaluated. Significant decreases (P < 0.05) of plasma vitamin C, vitamin E and carotenoids were found between T1 and T2 but not between T0 and T1. At T2, high plasma vitamin E was associated with high numbers of total and mature oocytes retrieved per patient, which, in turn, were favourable for achieving pregnancy. Accordingly, women who became pregnant presented higher vitamin E concentrations both in plasma and FF than those who did not. In conclusion, this study confirmed the occurrence of significant modifications of the plasma antioxidant profile during ovarian stimulation with gonadotrophins; at the same time, it was found that both systemic and follicular antioxidant status may be related to IVF/ICSI outcome.
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis