Merck
  • Differential thermodynamic driving force of first- and second-generation antihistamines to determine their binding affinity for human H1 receptors.
Differential thermodynamic driving force of first- and second-generation antihistamines to determine their binding affinity for human H1 receptors.
Biochemical pharmacology (2014-07-30)
Shigeru Hishinuma, Kenta Sugawara, Yoshihiro Uesawa, Hiroyuki Fukui, Masaru Shoji
ABSTRACT
Differential binding sites for first- and second-generation antihistamines were indicated on the basis of the crystal structure of human histamine H1 receptors. In this study, we evaluated differences between the thermodynamic driving forces of first- and second-generation antihistamines for human H1 receptors and their structural determinants. The binding enthalpy and entropy of 20 antihistamines were estimated with the van't Hoff equation using their dissociation constants obtained from their displacement curves against the binding of [(3)H]mepyramine to membrane preparations of Chinese hamster ovary cells expressing human H1 receptors at various temperatures from 4°C to 37°C. Structural determinants of antihistamines for their thermodynamic binding properties were assessed by quantitative structure-activity relationship (QSAR) analyses. We found that entropy-dependent binding was more evident in second- than first-generation antihistamines, resulting in enthalpy-entropy compensation between the binding forces of first- and second-generation antihistamines. QSAR analyses indicated that enthalpy-entropy compensation was determined by the sum of degrees, maximal electrostatic potentials, water-accessible surface area and hydrogen binding acceptor count of antihistamines to regulate their affinity for receptors. In conclusion, it was revealed that entropy-dependent hydrophobic interaction was more important in the binding of second-generation antihistamines, even though the hydrophilicity of second-generation antihistamines is generally increased. Furthermore, their structural determinants responsible for enthalpy-entropy compensation were explored by QSAR analyses. These findings may contribute to understanding the fundamental mechanisms of how the affinity of ligands for their receptors is regulated.
Sigma-Aldrich
Promethazine hydrochloride
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Chlorpheniramine maleate, United States Pharmacopeia (USP) Reference Standard
Promethazine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Betahistine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Desloratadine, powder, ≥98% (HPLC)
Supelco
Promethazine hydrochloride, vial of 25 mg, certified reference material, Cerilliant®
Promethazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Promethazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Desloratadine, European Pharmacopoeia (EP) Reference Standard
USP
Loratadine Related Compound A, United States Pharmacopeia (USP) Reference Standard
Supelco
Promethazine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Terfenadine
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Cyclizine hydrochloride, (European Pharmacopoeia (EP) Reference Standard)
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyrilamine maleate salt
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Betahistine dihydrochloride, ≥98%
Sigma-Aldrich
Oxatomide, ≥99%
Sigma-Aldrich
Fexofenadine hydrochloride, >98% (HPLC)
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Fexofenadine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Doxepin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Diphenhydramine hydrochloride, United States Pharmacopeia (USP) Reference Standard