Merck
  • Molecularly imprinted polymer as in-line concentrator in capillary electrophoresis coupled with mass spectrometry for the determination of quinolones in bovine milk samples.
Molecularly imprinted polymer as in-line concentrator in capillary electrophoresis coupled with mass spectrometry for the determination of quinolones in bovine milk samples.
Journal of chromatography. A (2014-08-16)
David Moreno-González, Francisco J Lara, Laura Gámiz-Gracia, Ana M García-Campaña
ABSTRACT
In this work molecularly imprinted polymers have been evaluated as sorbent for the construction of an in-line solid phase extraction analyte concentrator in capillary electrophoresis coupled with mass spectrometry for the determination of the eight regulated veterinary quinolones in bovine milk samples. Different parameters affecting the analyte concentrator performance, such as sample pH, volume and composition of the elution plug and injection time, were studied. Sample volumes of 22μL (2bar for 15min) were loaded on the MISPE microcartridge and the retained analytes were eluted by injecting a plug of MeOH/H2O/NH3 (60/37/3 by volume) for 125s at 50mbar (60nL). The proposed method is simple for the monitoring of these antibiotic residues in milk samples, allowing the direct injection of the samples with minimum sample pretreatment, achieving limits of detection between 3.8 and 4.7μgkg(-1) and unequivocal identification of the compounds working in tandem mass spectrometry. Recoveries ranging from 70.0 to 102.3% were obtained and satisfactory intra-day and inter-day RSDs were achieved (≤12% and 15% respectively). Reproducibility among different constructed analyte concentrators showed RSD≤11%.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%