Merck
  • Graphene oxide-based microspheres for the dispersive solid-phase extraction of non-steroidal estrogens from water samples.
Graphene oxide-based microspheres for the dispersive solid-phase extraction of non-steroidal estrogens from water samples.
Journal of chromatography. A (2014-12-03)
Yingying Wen, Zongliang Niu, Yanling Ma, Jiping Ma, Lingxin Chen
ABSTRACT
A modified Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) method based on the dispersive solid-phase extraction (dSPE) combined with high performance liquid chromatography (HPLC) was developed for the determination of non-steroidal estrogens in water samples. In this study, graphene oxide-based silica microspheres (SiO2@GO) were used as dSPE material for the preconcentration of analytes. HPLC was used for the separation and detection. This was the first time that the synthesized SiO2@GO microspheres were used as stationary phases for the off-line preconcentration of the non-steroidal estrogens in dSPE. dSPE parameters, such as sample pH, volume and type of eluent were optimized. Application of the developed method to analyze spiked lake, reservoir and tap water samples resulted in good recoveries values ranging from 70 to 106% with relative standard deviation values lower than 7.0% in all cases. Limits of detection were in the range of 0.2-6.1 μg/L. The combined data obtained in this study recommended that the proposed method is very fast, simple, repeatable and accurate for the detection of non-steroidal estrogens. Furthermore, the SiO2@GO microspheres application could potentially be expanded to extract and pre-concentrate other compounds in various matrices.
Product Description
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis