Merck
  • LC-MS/MS simultaneous quantitation of 2-hydroxyethylated, oxidative, and unmodified DNA nucleosides in DNA isolated from tissues of mice after exposure to ethylene oxide.
LC-MS/MS simultaneous quantitation of 2-hydroxyethylated, oxidative, and unmodified DNA nucleosides in DNA isolated from tissues of mice after exposure to ethylene oxide.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Fagen Zhang, Michael J Bartels, Matthew J LeBaron, Melissa R Schisler, Yo-Chan Jeong, B Bhaskar Gollapudi, Nigel P Moore
ABSTRACT
2-Hydroxyethylated and oxidative DNA nucleosides (DNA adduct biomarkers), such as O6-(2-hydroxyethyl)-2'-deoxyguanosine (O6HEdG), N6-(2-hydroxyethyl)-2'-deoxyadenosine (N6HEdA), 1-(2-hydroxyethyl)-2'-deoxyadenosine (N1HEdA), and 8-hydroxy-2'-deoxyguanosine (8-OHdG), N2,3-etheno-2'-deoxyguanosine (N2,3-ethenodG), α-methyl-γ-hydroxy-1,N2-propano-2'-deoxyguanosine (CrotondG), are important proposed biomarkers for exploring the genotoxicity mechanism of ethylene oxide (EO) in vivo. A liquid chromatography-tandem mass spectrometric method was developed for the simultaneous determination of O6HEdG, N6HEdA, N1HEdA, 8-OHdG, CrotondG, and N2,3-ethenodG together with regular 2'-deoxyguanosine (dG), and 2'-deoxyadenosine (dA) nucleosides in the DNA extracted from mouse lung tissues for the assessment of exposure to EO after inhalation. The lower limits of quantitation for 8-OHdG, CrotondG, N2,3-EthenodG, O6HEdG, N1HEdA, N6HEdA, dG, and dA were 0.025, 0.00125, 0.025, 0.00125, 0.025, 0.01, 2342, and 2500ng/mL, respectively. The linearity of the calibration curves for all analytes were >0.989. The intra-day assay precision relative standard deviation (RSD) values for quality control (QC) samples for all analytes were ≤13.5% with accuracy values ranging from 86.5% to 111%. The inter-day assay precision (RSD) values for all analytes were ≤18.8% with accuracy values ranging from 87.9% to 119%. This method was used for simultaneous determination of the levels of 8-OHdG, CrotondG, N2,3-EthenodG, O6HEdG, dG, N1HEdA, N6HEdA, and dA in DNA enzymatic hydrolysates from DNA extracted from mouse lung after 12 weeks' inhalation exposure to EO at atmospheric concentrations of 0, 100, and 200ppm. Overall, N2,3-ethenodG was not detected in any samples. 8-OHdG, CrotondG, dG, and dA were all quantifiable in all samples. O6HEdG, N1HEdA, and N6HEdA were quantifiable in most samples and the ratio of the corresponding adduct versus their corresponding DNA base (dG or dA) [×10 (e6)] was increased as the EO exposure concentration increased.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%