Merck
  • Flavonol glycosides and other phenolic compounds in buds and leaves of different varieties of black currant (Ribes nigrum L.) and changes during growing season.
Flavonol glycosides and other phenolic compounds in buds and leaves of different varieties of black currant (Ribes nigrum L.) and changes during growing season.
Food chemistry (2014-05-07)
Pengzhan Liu, Heikki Kallio, Baoru Yang
ABSTRACT
Phenolic compounds in buds and leaves of three varieties of black currant in Finland were identified by HPLC-DAD-ESI-MS/MS. Forty-three phenolic compounds of flavonol glycosides, proanthocyanidins and phenolic acids were found in variety "Mikael" whereas only thirty-five in "Mortti" and "Jaloste n:o 15". Glycosides of quercetin and kaempferol were the major phenolics. Rutin, hyperoside, isoquercitrin, kaempferol-3-O-rutinoise, kaempferol-3-O-glucoside, quercetin-3-O-(6″-malonyl)-glucoside and a kaempferol-malonylhexoside were the most abundant flavonol glycosides. The contents of flavonol glycosides ranged from 1 to 7 mg/g fresh weight in leaves showing typically an increasing trend from July to August, reaching the highest values in early October in "Mikael" and the end of August in "Mortti" and "Jaloste n:o 15". This is the first systematic report of the composition and content of phenolic compounds in buds and leaves of black currant.
Product Description
Supelco
Acetone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, natural, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Pricing and availability is not currently available.
Chlorogenic acid, primary reference standard
Pricing and availability is not currently available.
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Pricing and availability is not currently available.
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Ultrapure Acetonitrile
Pricing and availability is not currently available.
Supelco
Acetonitrile, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Pricing and availability is not currently available.