Merck
  • Association of single nucleotide polymorphisms in the gene encoding GLUT1 and diabetic nephropathy in Brazilian patients with type 1 diabetes mellitus.
Association of single nucleotide polymorphisms in the gene encoding GLUT1 and diabetic nephropathy in Brazilian patients with type 1 diabetes mellitus.
Clinica chimica acta; international journal of clinical chemistry (2015-02-24)
T Marques, T A Patente, M B Monteiro, A M Cavaleiro, M S Queiroz, M Nery, M J de Azevedo, L H Canani, M C Parisi, A Moura-Neto, M Passarelli, D Giannella-Neto, U F Machado, M L Corrêa-Giannella
ABSTRACT
Mesangial cells subject to high extracellular glucose concentrations, as occur in hyperglycaemic states, are unable to down regulate glucose influx, resulting in intracellular activation of deleterious biochemical pathways. A high expression of GLUT1 participates in the development of diabetic glomerulopathy. Variants in the gene encoding GLUT1 (SLC2A1) have been associated to this diabetic complication. The aim of this study was to test whether polymorphisms in SLC2A1 confer susceptibility to diabetic nephropathy (DN) in Brazilian type 1 diabetes patients. Four polymorphisms (rs3820589, rs1385129, rs841847 and rs841848) were genotyped in a Brazilian cohort comprised of 452 patients. A prospective analysis was performed in 155 patients. Mean duration of follow-up was 5.6 ± 2.4 years and the incidence of renal events was 18.0%. The rs3820589 presented an inverse association with the prevalence of incipient DN (OR: 0.36, 95% CI: 0.16 - 0.80, p=0.01) and with progression to renal events (HR: 0.20; 95% CI: 0.03 - 0.70; p=0.009). AGGT and AGAC haplotypes were associated with the prevalence of incipient DN and the AGAC haplotype was also associated with the prevalence of established/advanced DN. In conclusion, rs3820589 in the SLC2A1 gene modulates the risk to DN in Brazilian patients with inadequate type 1 diabetes control.
Product Description
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Benzethonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)
Benzethonium chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzethonium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material