Merck
  • A novel approach to the quantitative detection of anabolic steroids in bovine muscle tissue by means of a hybrid quadrupole time-of-flight-mass spectrometry instrument.
A novel approach to the quantitative detection of anabolic steroids in bovine muscle tissue by means of a hybrid quadrupole time-of-flight-mass spectrometry instrument.
Journal of chromatography. A (2014-08-19)
Julie Vanden Bussche, Anneleen Decloedt, Lieven Van Meulebroek, Nathalie De Clercq, Stephen Lock, Jianru Stahl-Zeng, Lynn Vanhaecke
ABSTRACT
In recent years, the analysis of veterinary drugs and growth-promoting agents has shifted from target-oriented procedures, mainly based on liquid chromatography coupled to triple-quadrupole mass spectrometry (LC-QqQ-MS), towards accurate mass full scan MS (such as Time-of-Flight (ToF) and Fourier Transform (FT)-MS). In this study, the performance of a hybrid analysis instrument (i.e. UHPLC-QuadrupoleTime-of-Flight-MS (QqToF-MS)), able to exploit both full scan HR and MS/MS capabilities within a single analytical platform, was evaluated for confirmatory analysis of anabolic steroids (gestagens, estrogens including stilbenes and androgens) in meat. The validation data was compared to previously obtained results (CD 2002/657/EC) for QqQ-MS and single stage Orbitrap-MS. Additionally, a fractional factorial design was used to shorten and optimize the sample extraction. Validation according to CD 2002/657/EC demonstrated that steroid analysis using QqToF has a higher competing value towards QqQ-MS in terms of selectivity/specificity, compared to single stage Orbitrap-MS. While providing excellent linearity, based on lack-of-fit calculations (F-test, α=0.05 for all steroids except 17β-ethinylestradiol: α=0.01), the sensitivity of QqToF-MS proved for 61.8% and 85.3% of the compounds more sensitive compared to QqQ-MS and Orbitrap-MS, respectively. Indeed, the CCα values, obtained upon ToF-MS/MS detection, ranged from 0.02 to 1.74μgkg(-1) for the 34 anabolic steroids, while for QqQ-MS and Orbitrap-MS values ranged from 0.04 to 0.88μgkg(-1) and from 0.07 to 2.50μgkg(-1), respectively. Using QqToF-MS and QqQ-MS, adequate precision was obtained as relative standard deviations for repeatability and within-laboratory reproducibility, were below 20%. In case of Orbitrap-MS, some compounds (i.e. some estrogens) displayed poor precision, which was possibly caused by some lack of sensitivity at lower concentrations and the absence of MRM-like experiments. Overall, it can be concluded that QqToF-MS offers good quantitative and confirmatory performance using the ToF-MS/MS mode whereas the full scan HR-ToF-MS allows screening for potential new designer drugs.
Product Description
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Norgestrel, European Pharmacopoeia (EP) Reference Standard
USP
Norgestrel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
USP
Medroxyprogesterone acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Methyltestosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Methyltestosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
17α-Methyltestosterone, ≥97.0% (HPLC)
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
17β-Hydroxy-17-methylandrosta-1,4-dien-3-one, analytical standard
Medroxyprogesterone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Dienestrol, European Pharmacopoeia (EP) Reference Standard