Merck
  • Increased sensitivity of apolipoprotein E knockout mice to copper-induced oxidative injury to the liver.
Increased sensitivity of apolipoprotein E knockout mice to copper-induced oxidative injury to the liver.
Biochemical and biophysical research communications (2015-03-10)
Yuan Chen, Bin Li, Ran-ran Zhao, Hui-feng Zhang, Chao Zhen, Li Guo
ABSTRACT
Apolipoprotein E (ApoE) genotypes are related to clinical presentations in patients with Wilson's disease, indicating that ApoE may play an important role in the disease. However, our understanding of the role of ApoE in Wilson's disease is limited. High copper concentration in Wilson's disease induces excessive generation of free oxygen radicals. Meanwhile, ApoE proteins possess antioxidant effects. We therefore determined whether copper-induced oxidative damage differ in the liver of wild-type and ApoE knockout (ApoE(-/-)) mice. Both wild-type and ApoE(-/-) mice were intragastrically administered with 0.2 mL of copper sulfate pentahydrate (200 mg/kg; a total dose of 4 mg/d) or the same volume of saline daily for 12 weeks, respectively. Copper and oxidative stress markers in the liver tissue and in the serum were assessed. Our results showed that, compared with the wild-type mice administered with copper, TBARS as a marker of lipid peroxidation, the expression of oxygenase-1 (HO-1), NAD(P)H dehydrogenase, and quinone 1 (NQO1) significantly increased in the ApoE(-/-) mice administered with copper, meanwhile superoxide dismutase (SOD) activity significantly decreased. Thus, it is concluded that ApoE may protect the liver from copper-induced oxidative damage in Wilson's disease.
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Xanthine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Xanthine, BioUltra, ≥99%