Merck
  • Structural and temperature effects on enantiomer separations of bicyclo[2.2.2]octane-based 3-amino-2-carboxylic acids on cinchona alkaloid-based zwitterionic chiral stationary phases.
Structural and temperature effects on enantiomer separations of bicyclo[2.2.2]octane-based 3-amino-2-carboxylic acids on cinchona alkaloid-based zwitterionic chiral stationary phases.
Journal of pharmaceutical and biomedical analysis (2014-06-09)
István Ilisz, Nóra Grecsó, Márta Palkó, Ferenc Fülöp, Wolfgang Lindner, Antal Péter
ABSTRACT
Procedures for the direct high-performance liquid chromatographic enantiomer separation of four bicyclo[2.2.2]octane-based 3-amino-2-carboxylic acids were developed in polar-ionic mode on zwitterionic chiral stationary phases (CSPs) based on cinchonane alkaloide quinine, quinidine and chiral sulfonic acid motifs. The effects of the mobile phase composition including the type of acid and base additives, the structures of the analytes and temperature were investigated. Experiments were performed at constant mobile phase compositions in the temperature range 10-50°C in order to study the effects of temperature, and thermodynamic parameters were calculated from plots of ln k or ln α vs. 1/T. Some mechanistic aspects of the chiral recognition process are discussed with respect to the structures of the analytes. It was found that the enantiomeric separations were in most cases enthalpically driven, but entropically driven separation was also observed. The sequence of elution of the enantiomers on the pseudo-enantiomerically behaving CSPs was determined in all cases.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Triethylamine, analytical standard
Supelco
Propylamine, analytical standard
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Ethylamine solution, 2.0 M in THF
Millipore
Bifido Selective Supplement B, suitable for microbiology