Merck
  • Neutrophils in antiretroviral therapy-controlled HIV demonstrate hyperactivation associated with a specific IL-17/IL-22 environment.
Neutrophils in antiretroviral therapy-controlled HIV demonstrate hyperactivation associated with a specific IL-17/IL-22 environment.
The Journal of allergy and clinical immunology (2014-07-22)
Laure Campillo-Gimenez, Sarah Casulli, Yasmine Dudoit, Sophie Seang, Guislaine Carcelain, Sidonie Lambert-Niclot, Victor Appay, Brigitte Autran, Roland Tubiana, Carole Elbim
ABSTRACT
Despite control of HIV infection under antiretroviral therapy (ART), immune T-cell activation persists in patients with controlled HIV infection, who are at higher risk of inflammatory diseases than the general population. PMNs play a key role in host defenses against invading microorganisms but also potentiate inflammatory reactions in cases of excessive or misdirected responses. The aim of our study was to analyze PMN functions in 60 ART-treated and controlled HIV-infected patients (viral load, <20 RNA copies/mL; CD4 count, ≥ 350 cells/mm(3)) with (HIV[I] group) and without (HIV[NI] group) diseases related to an inflammatory process and to compare them with 22 healthy control subjects. Flow cytometry was used to evaluate PMN functions in whole-blood conditions. We studied in parallel the activation markers of T lymphocytes and monocytes and the proinflammatory cytokine environment. Blood samples from HIV-infected patients revealed basal PMN hyperactivation associated with deregulation of the apoptosis/necrosis equilibrium. Interestingly, this hyperactivation was greater in HIV(I) than HIV(NI) patients and contrasted with a lack of monocyte activation in both groups. The percentage of circulating cells producing IL-17 was also significantly higher in HIV-infected patients than in control subjects and was positively correlated with markers of basal PMN activation. In addition, the detection of IL-22 overproduction in HIV(NI) patients suggests that it might contribute to counteracting chronic inflammatory processes during HIV infection. This study thus demonstrates the presence of highly activated PMNs in HIV-infected patients receiving effective ART and the association of these cells with a specific IL-17/IL-22 environment.
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Human CD14 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)