Merck
  • Wide injection zone compression in gradient reversed-phase liquid chromatography.
Wide injection zone compression in gradient reversed-phase liquid chromatography.
Journal of chromatography. A (2015-03-10)
Martin Gilar, Thomas S McDonald, Jay S Johnson, James P Murphy, James W Jorgenson
ABSTRACT
Chromatographic zone broadening is a common issue in microfluidic chromatography, where the sample volume introduced on column often exceeds the column void volume. To better understand the propagation of wide chromatographic zones on a separation device, a series of MS Excel spreadsheets were developed to simulate the process. To computationally simplify these simulations, we investigated the effects of injection related zone broadening and its gradient related zone compression by tracking only the movements of zone boundaries on column. The effects of sample volume, sample solvent, gradient slope, and column length on zone broadening were evaluated and compared to experiments performed on 0.32mm I.D. microfluidic columns. The repetitive injection method (RIM) was implemented to generate experimental chromatograms where large sample volume scenarios can be emulated by injecting two discrete small injection plugs spaced in time. A good match between predicted and experimental RIM chromatograms was observed. We discuss the performance of selected retention models on the accuracy of predictions and use the developed spreadsheets for illustration of gradient zone focusing for both small molecules and peptides.
Sigma-Aldrich
Butyrophenone, ≥99%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Indole, ≥99%, FG
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Hexanophenone, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Valerophenone, 99%
Supelco
Acetophenone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propiophenone, analytical standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Reserpine, crystallized, ≥99.0% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%