Merck
  • Multi-modal detection of colon malignancy by NIR-tagged recognition polymers and ultrasound contrast agents.
Multi-modal detection of colon malignancy by NIR-tagged recognition polymers and ultrasound contrast agents.
International journal of pharmaceutics (2014-12-02)
Meital Bloch, Lauren Jablonowski, Eylon Yavin, Dorit Moradov, Irena Djavsarov, Abraham Nyska, Margaret Wheatley, Abraham Rubinstein
ABSTRACT
To increase colonoscopy capability to discriminate benign from malignant polyps, we suggest combining two imaging approaches based on targeted polymeric platforms. Water-soluble cationized polyacrylamide (CPAA) was tagged with the near infrared (NIR) dye IR-783-S-Ph-COOH to form Flu-CPAA. The recognition peptide VRPMPLQ (reported to bind specifically to CRC tissues) was then conjugated with the Flu-CPAA to form Flu-CPAA-Pep which was then incorporated into echogenic microbubbles (MBs) made of polylactic acid (PLA) that are highly responsive to ultrasound. The ultimate design includes intravenous administration combined with local ultrasound and intra-colon inspection at the NIR range. In this proof of principle study PLA MBs were prepared by the double emulsion technique and loaded with several types of Flu-CPAA-Pep polymers. After insonation the submicron PLA fragments (SPF)-containing Flu-CPAA-Pep were examined in vitro for their ability to attach to colon cancer cells and in vivo (DMH induced rat model) for their ability to attach to colon malignant tissues and compared to the specific attachment of the free Flu-CPAA-Pep. The generation of SPF-containing Flu-CPAA-Pep resulted in a tissue attachment similar to that of the free, unloaded Flu-CPAA-Pep. The addition of VRPMPLQ to the polymeric backbone of the Flu-CPAA reduced cytotoxicity and improved the specific binding.
Product Description
Supelco
2,6-Lutidine, analytical standard
Sigma-Aldrich
2,6-Lutidine
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
2,6-Lutidine, ReagentPlus®, 98%
Sigma-Aldrich
Triethylsilane, 99%
Sigma-Aldrich
2,6-Dimethylpyridine, ≥99%
Sigma-Aldrich
Triethylsilane, 97%