Merck
Improved synthesis of chalcones under ultrasound irradiation.
Ultrasonics sonochemistry (2002-10-10)
Ji-Tai Li, Wen-Zhi Yang, Shu-Xiang Wang, Sheng-Hui Li, Tong-Shuang Li
ABSTRACT
Claisen-Schmidt condensation of acetophenone with aromatic aldehydes catalyzed by pulverized KOH and KF-Al2O3 results chalcones in 52-97% and 83-98% yields respectively in alcoholic solvent under ultrasound irradiation.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)