Merck
  • Analysis of early morbidity and functional outcome of thulium: yttrium-aluminum-garnet laser enucleation for benign prostate enlargement: patient age and prostate size determine adverse surgical outcome.
Analysis of early morbidity and functional outcome of thulium: yttrium-aluminum-garnet laser enucleation for benign prostate enlargement: patient age and prostate size determine adverse surgical outcome.
Urology (2014-12-23)
Steffen Rausch, Thomas Heider, Jens Bedke, Stephan Kruck, Christian Schwentner, Karsten Fischer, Arnulf Stenzl, Tilman Kälble
ABSTRACT
To evaluate complications and functional outcome and to identify patient-associated risk factors, we analyzed consecutive patients undergoing thulium:yttrium-aluminum-garnet laser enucleation of the prostate (ThuLEP) in our department. A total of 234 patients were prospectively analyzed. Preoperative data, postoperative complications, and outcome at 6, 12, and 24 months were recorded. Individual risk factors for complications and treatment failure were assessed by univariate and multivariate analyses. Mean age at surgery was 72.88 ± 7.83 years. Mean preoperative prostate size was 84.8 ± 34.9 mL. Thirty-day complication rate was 19.7%. Functional treatment failure occurred in 9.0% of all patients. Decline of mean International Prostate Symptom Score was -75%, quality of life index -76%, and postvoid residual -86% at 24 months. Maximum urine flow at 24 months was improved at +231%. In univariate analysis, age >80 years and prostate size <50 mL were significant predictors of complications, which was confirmed by multivariate analysis (P = .0277 and .0409, respectively). Age >80 years, prostate size <80 mL or <50 mL, and American Society of Anesthesiologists classification were significant predictors of functional treatment failure in univariate analysis. Prostate size <80 mL or <50 mL was significantly associated with treatment failure (P < .001) in multivariate analysis. ThuLEP is a safe and efficient surgical procedure, even in a patient cohort with high prostate volumes, age, and comorbidities. However, high patient age and small prostate size were significant determinants of adverse outcomes after surgery. To address the question of optimal therapy selection for patients with prostates smaller than 80 mL, further prospective randomized evaluation of ThuLEP and alternative surgical interventions is needed.
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, wire reel, 5m, diameter 2.0mm, as drawn, 99.999%
Aluminum, wire reel, 5m, diameter 3.0mm, as drawn, 99.999%
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, microleaf disks, 10mm, thinness 1.0μm, specific density 269.9μg/cm2, removable support, 99.999%
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, rod, 500mm, diameter 5.0mm, as drawn, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, rod, 100mm, diameter 12.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.05mm, as drawn, 99.99+%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, sphere, 100pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%