Merck
  • Evidence that collapsin response mediator protein-2 is involved in the dynamics of microtubules.
Evidence that collapsin response mediator protein-2 is involved in the dynamics of microtubules.
The Journal of biological chemistry (2000-04-20)
Y Gu, Y Ihara
ABSTRACT
Collapsin response mediator protein-2 (CRMP-2) is a member of the CRMP/TOAD/Ulip/DRP family of cytosolic phosphoproteins involved in neuronal differentiation and axonal guidance. CRMP-2 mediates the intracellular response to collapsin 1/semaphorin 3A, a repulsive extracellular guidance cue for axonal outgrowth. The mutation of UNC-33, a Caenorhabditis elegans homolog of CRMP-2, results in abnormality of microtubules in neurites, but the mechanism of CRMP-2 action remains to be clarified. Here, we report that overexpression of human CRMP-2 in Neuro2a cells, a mouse neuroblastoma cell line, results in blebbing of the cytoplasm. Furthermore, some cells exhibited intranuclear inclusions, which were labeled with antibodies to CRMP-2 and tubulin. CRMP-2 was found to be associated with microtubule bundles in the spindles at the metaphase and in the midbodies at the late telophase in mitotic cells. Thus, it is most likely that failure of complete disassembly of the spindle microtubules during mitosis is responsible for the formation of these intranuclear inclusions. We suggest that CRMP-2 functions by regulating the dynamics of microtubules.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid