Merck
  • Polyethylenimine600-β-cyclodextrin: a promising nanopolymer for nonviral gene delivery of primary mesenchymal stem cells.
Polyethylenimine600-β-cyclodextrin: a promising nanopolymer for nonviral gene delivery of primary mesenchymal stem cells.
International journal of nanomedicine (2013-06-06)
Haijun Tong, Chuandong Wang, Yan Huang, Qin Shi, Julio C Fernandes, Kerong Dai, Guping Tang, Xiaoling Zhang
ABSTRACT
Genetically modified mesenchymal stem cells (MSCs) have great potential in the application of regenerative medicine and molecular therapy. In the present manuscript, we introduce a nanopolymer, polyethylenimine600-β-cyclodextrin (PEI600-β-CyD), as an efficient polyplex-forming plasmid delivery agent with low toxicity and ideal transfection efficiency on primary MSCs. PEI600-β-CyD causes significantly less cytotoxicity and apoptosis on MSCs than 25 kDa high-molecular-weight PEI (PEI25kDa). PEI600-β-CyD also exhibits similar transfection efficiency as PEI25kDa on MSCs, which is higher than that of PEI600Da. Quantum dot-labeled plasmids show that PEI600-β-CyD or PEI25kDa delivers the plasmids in a more scattered manner than PEI600Da does. Further study shows that PEI600-β-CyD and PEI25kDa are more capable of delivering plasmids into the cell lysosome and nucleus than PEI600Da, which correlates well with the results of their transfection-efficiency assay. Moreover, among the three vectors, PEI600-β-CyD has the most capacity of enhancing the alkaline phosphatase activity of MSCs by transfecting bone morphogenetic protein 2, 7, or special AT-rich sequence-binding protein 2. These results clearly indicate that PEI600-β-CyD is a safe and effective candidate for the nonviral gene delivery of MSCs because of its ideal inclusion ability and proton sponge effect, and the application of this nanopolymer warrants further investigation.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture