Merck
  • Analysis of the band profiles of the enantiomers of phenylglycine in liquid chromatography on bonded teicoplanin columns using the stochastic theory of chromatography.
Analysis of the band profiles of the enantiomers of phenylglycine in liquid chromatography on bonded teicoplanin columns using the stochastic theory of chromatography.
Journal of chromatography. A (2001-07-19)
P Jander, V Backovská, A Felinger
ABSTRACT
The retention behaviour of the enantiomers of underivatized phenylglycine was studied on a Chirobiotic T column packed with amphoteric glycopeptide teicoplanin covalently bonded to the surface of silica gel. The retention and the selectivity of separation of the enantiomers increase with rising concentration of ethanol or of methanol in aqueous-organic mobile phases. The band profiles of the less retained L-phenylglycine are symmetrical, but the band profiles of the more strongly retained D-phenylglycine are tailing in all mobile phases tested. The band broadening does not diminish even at very low concentrations of phenylglycine, so that it cannot be attributed to possible column overload. The analysis of the band profile using the stochastic theory of chromatography suggests that the broadening can be attributed to at least two additional chiral centres of adsorption in the stationary phase contributing to the retention of the more strongly retained enantiomer in addition to the adsorption of the less retained one. This behaviour can be explained by the complex structure of the teicoplanin chiral stationary phase.
Product Description
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm